Learn More
Cytokeratin 20 Recombinant Rabbit Monoclonal Antibody (RM283), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Supplier: Thermo Scientific MA527906
Description
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Cytokeratin 20 Recombinant Monoclonal antibody specifically detects Cytokeratin 20 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
Cytokeratin 20 | |
Recombinant Monoclonal | |
0.05 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide | |
P35900 | |
KRT20 | |
A peptide corresponding to the C-terminus of human Cytokeratin-20 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
RM283 | |
Unconjugated | |
KRT20 | |
9030623C06Rik; CD20; CK20; CK-20; CK-21; cytokeratin 20; cytokeratin 21; cytokeratin-20; Cytokeratin-21; K20; keratin 20; keratin 20, type I; keratin, type I cytoskeletal 20; Keratin-20; krt20; Krt21; MGC35423; Protein IT | |
Rabbit | |
Protein A | |
RUO | |
54474 | |
-20°C | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.