Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 20 Rabbit anti-Human, Mouse, Non-human Primate, Polyclonal, FabGennix

Catalog No. 502206534
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-6534 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-6534 Supplier Fabgennix Inc Supplier No. CK20101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cytokeratin 20 Polyclonal antibody specifically detects Cytokeratin 20 in Human, Mouse, Non-human Primate samples. It is validated for ELISA, Immunoprecipitation, Western Blot

Specifications

Antigen Cytokeratin 20
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene KRT20
Gene Accession No. P35900, Q9D312
Gene Alias 9030623C06Rik; CD20; CK20; CK-20; CK-21; cytokeratin 20; cytokeratin 21; cytokeratin-20; Cytokeratin-21; K20; keratin 20; keratin 20, type I; keratin, type I cytoskeletal 20; Keratin-20; krt20; Krt21; MGC35423; Protein IT
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 300-350 human keratin, type I cytoskeletal 20 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54474, 66809, 744139
Target Species Human, Mouse, Non-human Primate
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.