Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytokeratin 23 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550198
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550198 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550198 Supplier Invitrogen™ Supplier No. PA550198
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total KRT23 protein.

The protein encoded by this gene is a member of the keratin family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. The type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains. The type I cytokeratin genes are clustered in a region of chromosome 17q12-q21.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 23
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene KRT23
Gene Accession No. Q9C075
Gene Alias C HAIK1; CK23; CK-23; cytokeratin 23; cytokeratin-23; DKFZP434G032; HAIK1; histone deacetylase inducible keratin 23; hyperacetylation-inducible type I keratin; K23; Ka23; keratin 23; keratin 23 (histone deacetylase inducible); keratin 23, type I; keratin complex 1, acidic, gene 23; keratin, type I cytoskeletal 23; Keratin-23; Krt1-23; KRT23; MGC26158; type I intermediate filament cytokeratin; type I keratin KA23
Gene Symbols KRT23
Host Species Rabbit
Immunogen Fusion protein corresponding to residues near the N terminal of human keratin 23 (histone deacetylase inducible).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25984
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.