Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytokeratin 26 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546514
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546514 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546514 Supplier Invitrogen™ Supplier No. PA546514
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: IDIYCNLLDG EERKSKSTCY KSKGYRPVNS GNQAKDSTEE TIVKTVVEEL Sequence homology: Human: 100%; Rabbit: 77%.

The protein encoded by this gene is a member of the keratin superfamily. This keratin protein is a type I keratin that is specific for the inner root sheath of the hair follicle. This gene exists in a cluster with other keratin genes on chromosome 17q21.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 26
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene KRT26
Gene Accession No. Q7Z3Y9
Gene Alias CK26; CK-26; Cytokeratin-26; K25; K25B; K25IRS2; K26; keratin 25B; keratin 26; keratin 26, type I; keratin, type I cytoskeletal 26; Keratin-25B; keratin-26; KRT25B; KRT26; type I inner root sheath specific keratin 25 irs2; type I inner root sheath-specific keratin-K25irs2
Gene Symbols KRT26
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human KRT26 (aa 384-433).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 353288
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.