Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytokeratin 27 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565924
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565924 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565924 Supplier Invitrogen™ Supplier No. PA565924
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RRLGSCGGTGS VRLSSGGAGF GAGNTCGVPG IGSGFS Highest antigen sequence identity to the following orthologs - mouse 65%, rat 68%.

Essential for the proper assembly of type I and type II keratin protein complexes and formation of keratin intermediate filaments in the inner root sheath (irs).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 27
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KRT27
Gene Accession No. Q7Z3Y8
Gene Alias CK-27; cytokeratin-27; K25C; K25IRS3; K27; keratin 25C; keratin 27; keratin 27, type I; keratin, type I cytoskeletal 27; Keratin-25C; keratin-27; KRT25C; KRT27; type I inner root sheath specific keratin 25 irs3; type I inner root sheath-specific keratin-K25irs3
Gene Symbols KRT27
Host Species Rabbit
Immunogen Recombinant Human Cytokeratin 27. Recombinant protein control fragment (Product #RP-106602).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 342574
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.