Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytokeratin 2e Monoclonal Antibody (D2), Invitrogen™

Catalog No. FEMA551052
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA551052 20 μL
FEMA551051 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEMA551052 Supplier Invitrogen™ Supplier No. MA551052
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytokeratins are a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. They range in molecular weight between 40 kDa and 68 kDa and isoelectric pH between 4.9 - 7.8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 2e
Applications Western Blot, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone D2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene KRT2
Gene Accession No. P35908, Q6IG02
Gene Alias BB005427; CK 2e; CK-2e; Cytokeratin-2e; epithelial keratin-2e; K2e; Kb2; keratin 2; keratin 2 epidermis; keratin 2, type II; keratin 2e; keratin complex 2, basic, gene 17; keratin complex 2, gene 17; keratin, type II cytoskeletal 2 epidermal; Keratin-2 epidermis; keratin-2e; KRT2; krt2 {ECO:0000250; Krt2-17; Krt2-2; KRT2A; Krt2e; KRTE; MGC116967; MGC116968; type II keratin Kb2; type-II keratin Kb2; UniProtKB:P35908}
Gene Symbols KRT2
Host Species Mouse
Immunogen Recombinant Human CK2 protein, Cys7-Leu160 (Accession #P35908).
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3849, 406228
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.