Learn More
Invitrogen™ Cytokeratin 2e Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587851
Description
Immunogen sequence: MSCQISCKSR GRGGGGGGFR GFSSGSAVVS GGSRRSTSSF SCLSRHGGGG GGFGGGGFGS RSLVGLGGTK SISISVAGGG GGFGAAGGFG GRGGGFGGGS SFGGGSGFSG GGFGGGGFGG GRFGGFGGPG GVGGLGGPGG FGPGGYPGGI HEVSVNQSLL QPLNVKVDPE IQNVKAQERE QIKTLNNKFA SFIDKVRFLE QQNQVLQTKW ELLQQMNVGT; Positive Samples: HeLa, Mouse liver.
Cytokeratins are a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. They range in molecular weight between 40 kDa and 68 kDa and isoelectric pH between 4.9 - 7.8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium.
Specifications
Cytokeratin 2e | |
Polyclonal | |
Unconjugated | |
KRT2 | |
BB005427; CK 2e; CK-2e; Cytokeratin-2e; epithelial keratin-2e; K2e; Kb2; keratin 2; keratin 2 epidermis; keratin 2, type II; keratin 2e; keratin complex 2, basic, gene 17; keratin complex 2, gene 17; keratin, type II cytoskeletal 2 epidermal; Keratin-2 epidermis; keratin-2e; KRT2; krt2 {ECO:0000250; Krt2-17; Krt2-2; KRT2A; Krt2e; KRTE; MGC116967; MGC116968; type II keratin Kb2; type-II keratin Kb2; UniProtKB:P35908} | |
Rabbit | |
Affinity Chromatography | |
RUO | |
16681, 3849 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
4.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P35908, Q3TTY5 | |
KRT2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human Cytokeratin 2e (KRT2) (NP_0004142). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.