Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 2e Rabbit anti-Human, Clone: 2F7, Bioss
SDP

Catalog No. 501989558
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9558 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9558 Supplier Bioss Supplier No. BSM52061R
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Cytokeratins are a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. They range in molecular weight between 40 kDa and 68 kDa and isoelectric pH between 4.9 - 7.8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium.

Specifications

Antigen Cytokeratin 2e
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2F7
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 40% glycerol and 0.05% sodium azide; pH
Gene KRT2
Gene Accession No. P35908
Gene Alias BB005427; CK 2e; CK-2e; Cytokeratin-2e; Epithelial keratin-2e; K2e; Kb2; keratin 2; keratin 2 epidermis; keratin 2, type II; keratin complex 2, basic, gene 17; keratin complex 2, gene 17; Keratin, type II cytoskeletal 2 epidermal; Keratin-2 epidermis; Keratin-2e; KRT2; krt2 {ECO:0000250; Krt2-17; Krt2-2; Krt2a; KRT2E; KRTE; MGC116967; MGC116968; type II keratin Kb2; Type-II keratin Kb2; UniProtKB:P35908}
Gene Symbols KRT2
Host Species Rabbit
Immunogen Recombinant human Cytokeratin 2e protein, around 350-500aa.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3849
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.