Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 3 Antibody (KRT3/2130) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279852
Description
Cytokeratin 3 Monoclonal specifically detects Cytokeratin 3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
Cytokeratin 3 | |
Monoclonal | |
1 mg/mL | |
Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
CK3, CK-3, cytokeratin-3, FLJ95909, K3cytokeratin 3, keratin 3, keratin, type II cytoskeletal 3,65 kDa cytokeratin, Keratin-3, Type-II keratin Kb3 | |
Mouse | |
Protein A or G purified | |
RUO | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
KRT3/2130 | |
Unconjugated | |
10 mM PBS with No Preservative | |
KRT3 | |
Recombinant full-length human Cytokeratin 3 protein (Uniprot: P12035) | |
100 μg | |
Primary | |
Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction