Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 3 Antibody (KRT3/2542), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308582AF405
Description
Cytokeratin 3 Monoclonal antibody specifically detects Cytokeratin 3 in Human, Bovine, Rabbit samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
Cytokeratin 3 | |
Monoclonal | |
Alexa Fluor 405 | |
CK3, CK-3, cytokeratin-3, FLJ95909, K3cytokeratin 3, keratin 3, keratin, type II cytoskeletal 3,65 kDa cytokeratin, Keratin-3, Type-II keratin Kb3 | |
Tissue, cells or virus corresponding to Rabbit Cytokeratin 3/CK-3. Rabbit corneal epithelial keratin. | |
100 μg | |
Cell Biology, Cytoskeleton Markers, Immunology, Neuroscience, Signal Transduction | |
3850 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, Immunohistochemistry (Paraffin) | |
KRT3/2542 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Bovine, Rabbit | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction