Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytokeratin 3 Monoclonal Antibody (AE5), FITC
GREENER_CHOICE

Catalog No. PIMA546557
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546557 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA546557 Supplier Invitrogen™ Supplier No. MA546557
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Predicted molecular weight: 65 kDa.

Cytokeratin 3 is a member of the keratin gene family. The type II cytokeratins consist of basic or neutral proteins which are arranged in pairs of heterotypic keratin chains coexpressed during differentiation of simple and stratified epithelial tissues. This type II cytokeratin is specifically expressed in the corneal epithelium with family member KRT12 and mutations in these genes have been associated with Meesmann's Corneal Dystrophy. The type II cytokeratins are clustered in a region of chromosome 12q12-q13. (NCBI Gene description).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 3
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Monoclonal
Clone AE5
Concentration 1 mg/mL
Conjugate FITC
Formulation PBS with 0.09% sodium azide
Gene KRT3
Gene Accession No. P12035, Q29426
Gene Alias 65 kDa cytokeratin; CK3; CK-3; cytokeratin 3; cytokeratin type II, component Ib/c; cytokeratin-3; K3; K3 keratin; keratin 3; keratin 3, type II; keratin K3; keratin, type II cytoskeletal 3; Keratin-3; KERIB; KRT3; Type-II keratin Kb3
Gene Symbols KRT3
Host Species Mouse
Immunogen Rabbit corneal epithelial keratins.
Purification Method Protein G
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100353668, 3850, 407188
Target Species Human, Rabbit, Bovine
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.