Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Cytokeratin 3 Polyclonal Antibody
Rabbit Polyclonal Antibody
$178.40 - $484.00
Specifications
Antigen | Cytokeratin 3 |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119244
|
Invitrogen™
PA5119244 |
200 μL |
Each for $484.00
|
|
|||||
PIPA5122291
|
Invitrogen™
PA5122291 |
20 μL |
Each for $178.40
|
|
|||||
Description
Cytokeratin 3 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Cytokeratin 3 is a member of the keratin gene family. The type II cytokeratins consist of basic or neutral proteins which are arranged in pairs of heterotypic keratin chains coexpressed during differentiation of simple and stratified epithelial tissues. This type II cytokeratin is specifically expressed in the corneal epithelium with family member KRT12 and mutations in these genes have been associated with Meesmann's Corneal Dystrophy. The type II cytokeratins are clustered in a region of chromosome 12q12-q13. (NCBI Gene description).Specifications
Cytokeratin 3 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human, Rat | |
KRT3 | |
65 kDa cytokeratin; CK3; CK-3; cytokeratin 3; cytokeratin type II, component Ib/c; cytokeratin-3; K3; K3 keratin; keratin 3; keratin 3, type II; keratin K3; keratin, type II cytoskeletal 3; Keratin-3; KERIB; KRT3; Type-II keratin Kb3 | |
KRT3 | |
Primary | |
Antigen affinity chromatography, Protein A |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
P12035 | |
3850 | |
Recombinant protein Keratin 3. The antigen corresponds to amino acid range Glu91-Lys394 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title