Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Cytokeratin 31 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562005
Description
Immunogen sequence: NACSKPIGPC LSNPCTSCVP PAPCTPCA Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 79%.
The protein encoded by this gene is a member of the keratin gene family. As a type I hair keratin, it is an acidic protein which heterodimerizes with type II keratins to form hair and nails. The type I hair keratins are clustered in a region of chromosome 17q12-q21 and have the same direction of transcription.
Specifications
Cytokeratin 31 | |
Polyclonal | |
Unconjugated | |
KRT31 | |
Ha1; Ha-1; hair keratin; Hair keratin, type I Ha1; hard keratin, type I, 1; hHa1; HKA1; HKA-1; K31; Ka25; keratin 31; keratin 31, type I; keratin complex 1, acidic, gene 1; keratin complex 1, gene 1; keratin, hair, acidic,1; keratin, type I cuticular Ha1; Keratin-31; Kha1; KRT hHa1; Krt1-1; Krt31; KRTHA1; MKHA-1; type I hair keratin KA25 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3881 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q15323 | |
KRT31 | |
Recombinant protein corresponding to Human Cytokeratin 31. Recombinant protein control fragment (Product #RP-101130). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction