Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 33A/33B Polyclonal Antibody, Invitrogen™

Catalog No. PIPA5103455
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103455 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103455 Supplier Thermo Scientific Supplier No. PA5103455

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total K1HB.

The protein encoded by this gene is a member of the keratin gene family. It is one of the type I hair keratin genes which are clustered in a region of chromosome 17q12-q21 and have the same direction of transcription. As a type I hair keratin, it is an acidic protein which heterodimerizes with type II keratins to form hair and nails. There are two isoforms of this protein, encoded by two separate genes, KRTHA3A and KRTHA3B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 33A/33B
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene KRT33B
Gene Accession No. Q14525
Gene Alias HA3II; Ha-3II; hair keratin; Hair keratin, type I Ha3-II; hard keratin, type I, 3II; hHa3-II; HKA3B; K33B; keratin 33B; keratin 33B, type I; keratin, hair, acidic, 3B; keratin, type I cuticular Ha3-II; Keratin-33B; KRT33B; KRTHA3A; KRTHA3B; type I hair keratin 3B
Gene Symbols KRT33B
Host Species Rabbit
Immunogen A synthesized peptide derived from human KRT33B(Accession Q14525), corresponding to amino acid residues L360-Y404.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3884
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.