Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytokeratin 33B Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA522237

Catalog No. PIPA522237


Only null left
Add to Cart

Description

Description

Recommended positive controls: HeLa. Predicted reactivity: Mouse (83%), Rat (86%), Dog (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is a member of the keratin gene family. It is one of the type I hair keratin genes which are clustered in a region of chromosome 17q12-q21 and have the same direction of transcription. As a type I hair keratin, it is an acidic protein which heterodimerizes with type II keratins to form hair and nails. There are two isoforms of this protein, encoded by two separate genes, KRTHA3A and KRTHA3B.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cytokeratin 33B
Polyclonal
Unconjugated
KRT33B
HA3II; Ha-3II; hair keratin; Hair keratin, type I Ha3-II; hard keratin, type I, 3II; hHa3-II; HKA3B; K33B; keratin 33B; keratin 33B, type I; keratin, hair, acidic, 3B; keratin, type I cuticular Ha3-II; Keratin-33B; KRT33B; KRTHA3A; KRTHA3B; type I hair keratin 3B
Rabbit
Antigen affinity chromatography
RUO
3884
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.44 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q14525
KRT33B
Recombinant protein encompassing a sequence within the center region of human Cytokeratin 33B. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.