Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 33B Polyclonal Antibody
Cytokeratin 33B Polyclonal Antibody
Supplier: Thermo Scientific PA598437
Description
Cytokeratin 33B Polyclonal Antibody for Western Blot, IHC (P), ELISA

Specifications
Cytokeratin 33B | |
Polyclonal | |
Unconjugated | |
KRT33B | |
HA3II; Ha-3II; hair keratin; Hair keratin, type I Ha3-II; hard keratin, type I, 3II; hHa3-II; HKA3B; K33B; keratin 33B; keratin 33B, type I; keratin, hair, acidic, 3B; keratin, type I cuticular Ha3-II; Keratin-33B; KRT33B; KRTHA3A; KRTHA3B; type I hair keratin 3B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3884 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
3 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q14525 | |
KRT33B | |
Recombinant Human Keratin, type I cuticular Ha3-II protein (1-404AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction