Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytokeratin 36 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531829
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531829 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531829 Supplier Invitrogen™ Supplier No. PA531829
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa. Predicted reactivity: Mouse (80%), Rat (82%), Bovine (83%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is a member of the keratin gene family. This type I hair keratin is an acidic protein which heterodimerizes with type II keratins to form hair and nails. The type I hair keratins are clustered in a region of chromosome 17q12-q21 and have the same direction of transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 36
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene KRT36
Gene Accession No. O76013
Gene Alias HA6; hair keratin; Hair keratin, type I Ha6; hHa6; HKA6; Hra-1; K36; Ka31; keratin 36; keratin 36, type I; keratin complex 1, acidic, gene 5; keratin, hair, acidic, 6; keratin, type I cuticular Ha6; Keratin-36; keratin-5; KRT hHa6; Krt1-22; Krt1-5; Krt36; KRTHA6; MHRa-1; type I hair keratin KA31; Type I keratin 48 kDa; type I keratin 48 kDa (MHRa-1)
Gene Symbols KRT36
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Keratin 36. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8689
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.