Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytokeratin 35/37/38 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103454
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103454 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103454 Supplier Invitrogen™ Supplier No. PA5103454
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total K1H7.

Members of the keratin gene family. This type I hair keratin is an acidic protein which heterodimerizes with type II keratins to form hair and nails. The type I hair keratins are clustered in a region of chromosome 17q12-q21 and have the same direction of transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 35/37/38
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene KRT37
Gene Accession No. O76014, O76015, Q92764
Gene Alias HA7; HA8; hair keratin; hair keratin, type I Ha7; Hair keratin, type I Ha8; HHA7; HHA8; HKA7; HKA8; K37; K38; keratin 37; keratin 37, type I; keratin 38; keratin 38, type I; keratin, hair, acidic, 7; keratin, hair, acidic, 8; keratin, type I cuticular Ha7; Keratin, type I cuticular Ha8; keratin-37; Keratin-38; KRT hHa8; KRT37; KRT38; KRTHA7; KRTHA8
Gene Symbols KRT35, KRT37, KRT38
Host Species Rabbit
Immunogen A synthesized peptide derived from human KRT37(Accession O76014), corresponding to amino acid residues C413-R449.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3886, 8687, 8688
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.