Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytokeratin 4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529822
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529822 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529822 Supplier Invitrogen™ Supplier No. PA529822
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: H1299, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (84%), Rat (85%), Dog (91%), Bovine (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Cytokeratins are a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. They range in molecular weight between approximately40 kDa and approximately68 kDa and isoelectric pH between 4. 9-7. 8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.35 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene KRT4
Gene Accession No. P19013
Gene Alias 57kd keratin; 57kDa keratin; AW108092; cb59; CK4; CK-4; CYK4; cytokeratin; cytokeratin 4; cytokeratin-4; Cytoskeletal 57 kDa keratin; etID20321.6; intermediate filament protein; K4; Kb4; keratin 4; keratin 4, type II; keratin 8; keratin complex 2, basic, gene 4; keratin, type II cytoskeletal 4; Keratin-4; Krt-2.4; Krt2-4; Krt4; krt4 {ECO:0000250; krt4 protein; krt8; type II basic cytokeratin; type II keratin Kb4; type-II keratin Kb4; UniProtKB:P19013}; WSN1; wu:fa91a11; wu:fd19e09; wu:fl23c09; zf-K8
Gene Symbols KRT4
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Cytokeratin 4. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3851
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.