Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytokeratin 4 Recombinant Rabbit Monoclonal Antibody (SN74-03)
GREENER_CHOICE

Catalog No. PIMA532319
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532319 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532319 Supplier Invitrogen™ Supplier No. MA532319
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Cytokeratins are a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. They range in molecular weight between approximately40 kDa and approximately68 kDa and isoelectric pH between 4. 9-7. 8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 4
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Recombinant Monoclonal
Clone SN74-03
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene KRT4
Gene Accession No. P19013
Gene Alias 57kd keratin; 57kDa keratin; AW108092; cb59; CK4; CK-4; CYK4; cytokeratin; cytokeratin 4; cytokeratin-4; Cytoskeletal 57 kDa keratin; etID20321.6; intermediate filament protein; K4; Kb4; keratin 4; keratin 4, type II; keratin 8; keratin complex 2, basic, gene 4; keratin, type II cytoskeletal 4; Keratin-4; Krt-2.4; Krt2-4; Krt4; krt4 {ECO:0000250; krt4 protein; krt8; type II basic cytokeratin; type II keratin Kb4; type-II keratin Kb4; UniProtKB:P19013}; WSN1; wu:fa91a11; wu:fd19e09; wu:fl23c09; zf-K8
Gene Symbols KRT4
Host Species Rabbit
Immunogen Recombinant protein within Human Cytokeratin 4 aa 395-520.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3851
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.