Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 5/6/18 Mouse anti-Human, Rat, Bovine, Clone: LP34, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00
Specifications
Antigen | Cytokeratin 5/6/18 |
---|---|
Clone | LP34 |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 0.1-0.2 ug/ml |
Applications | Immunohistochemistry (Paraffin) |
Description
Cytokeratin 5/6/18 Monoclonal specifically detects Cytokeratin 5/6/18 in Human, Rat, Bovine samples. It is validated for Immunohistochemistry-Paraffin.Specifications
Cytokeratin 5/6/18 | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cell Biology, Cellular Markers | |
10 mM PBS with 0.05% BSA | |
3852 | |
IgG1 κ | |
Protein A or G purified |
LP34 | |
Immunohistochemistry-Paraffin 0.1-0.2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human, Rat, Bovine | |
58 kDa cytokeratin, CK5, CK-5, cytokeratin-5, DDD, EBS2, epidermolysis bullosa simplex 2 Dowling-Meara/Kobner/Weber-Cockayne types, K5, keratin 5, keratin 5 (epidermolysis bullosa simplex, Dowling-Meara/Kobner/Weber-Cockaynetypes), keratin, type II cytoskeletal 5, Keratin-5, KRT5A, Type-II keratin Kb5 | |
Detergent-insoluble fraction of psoriatic human epidermis | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title