Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Cytokeratin 5/6 Ab-2, Mouse Monoclonal Antibody

Catalog No. MS1814S Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1814S 1 mL Concentrated
MS1814S0 100 μL Concentrated
MS1814S1 500 μL Concentrated
MS1814R7 7 mL Prediluted
MS1814RQ 12 mL Ready-to-Use
MS1814R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MS1814S Supplier Epredia™ Supplier No. MS1814S
Add to Cart
Add to Cart

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Cytokeratin 5/6 Ab-2, Mouse Monoclonal Antibody.

Twenty human keratins are divided into acidic (pI <5.7) and basic (pI >6.0) subfamilies. Members of the acidic and basic subfamilies are found together in pairs. The composition of keratin pairs varies with the epithelial cell type, stage of differentiation, cellular growth environment, and disease state. Many studies have shown the usefulness of keratins as markers in cancer research and tumor identification.

Host Species: Mouse

Clone: D5/16 B4

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Isolated cytokeratin 5

Molecular Weight: 58 kDa (Cytokeratin 5), 56 kDa (Cytokeratin 6)

Positive Control: Mesothelioma, Squamous Cell Ca

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Cytokeratin 5/6 Ab-2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone D5/16 B4
Conjugate Unconjugated
Dilution 1:20 for 30 min at RT
Host Species Mouse
Immunogen Isolated cytokeratin 5
Quantity 1 mL
Regulatory Status IVD
Research Discipline Cytoskeletal Protein
Primary or Secondary Primary
Target Species Human
Content And Storage Store vial at 4deg.C. When stored at 2 to 8deg.C,
Form Concentrated
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.