Learn More
Description
Host Species: Mouse
Clone: D5/16 B4
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Isolated cytokeratin 5
Molecular Weight: 58 kDa (Cytokeratin 5), 56 kDa (Cytokeratin 6)
Positive Control: Mesothelioma, Squamous Cell Ca
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | Cytokeratin 5/6 Ab-2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | D5/16 B4 |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Isolated cytokeratin 5 |
| Quantity | 500 μL |
| Regulatory Status | IVD |
| Research Discipline | Cytoskeletal Protein |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.