Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 5/6 Mouse anti-Human, Clone: KRT5.6/4866, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00
Specifications
Antigen | Cytokeratin 5/6 |
---|---|
Clone | KRT5.6/4866 |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
Applications | Immunohistochemistry (Paraffin) |
Description
Cytokeratin 5/6 Monoclonal specifically detects Cytokeratin 5/6 in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
Cytokeratin 5/6 | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cell Biology, Cellular Markers | |
10 mM PBS with 0.05% BSA | |
3852 | |
IgG1 κ, IgG2a κ | |
Protein A or G purified |
KRT5.6/4866 | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
58 kDa cytokeratin, CK5, CK-5, cytokeratin-5, DDD, EBS2, epidermolysis bullosa simplex 2 Dowling-Meara/Kobner/Weber-Cockayne types, K5, keratin 5, keratin 5 (epidermolysis bullosa simplex, Dowling-Meara/Kobner/Weber-Cockaynetypes), keratin, type II cytoskeletal 5, Keratin-5, KRT5A, Type-II keratin Kb5 | |
Recombinant full-length human Cytokeratin 5/6 protein | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title