Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 5/8 Antibody (RCK102), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197730AF405
Description
Cytokeratin 5/8 Monoclonal specifically detects Cytokeratin 5/8 in Human, Mouse, Rat, Porcine, Canine, Feline, Hamster, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Cytokeratin 5/8 | |
Monoclonal | |
Alexa Fluor 405 | |
cell proliferation-inducing protein 46, CK18, CK5, CYK18, DDD, K18, K1CO, K5, Keratin, Keratin 18, Keratin 5, KRT18, krt5, KRT5A, KRTB, type I cytoskeletal 18, type II cytoskeletal 5 | |
Mouse | |
Affinity Purified | |
RUO | |
3852 | |
Human, Mouse, Rat, Pig, Canine, Feline, Hamster, Rabbit | |
IgG1 |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
RCK102 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
KRT5 | |
Clone RCK102 is a mouse monoclonal IgG1 antibody derived by fusion of SP2/0 mouse myeloma cells with spleen cells from a mouse immunized with cytokeratins from a human lung cancer cell line (MR21). | |
0.1 mL | |
Primary | |
RCK102 is reacting with cytokeratin 5 and cytokeratin 8. This monoclonal antibody recognizes virtually all epithelial tissues and carcinomas. | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction