Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytokeratin 5/8 Monoclonal Antibody (RCK102)
GREENER_CHOICE

Catalog No. MA106314
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA106314 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA106314 Supplier Invitrogen™ Supplier No. MA106314
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-06314 detects cytokeratin 5 and 8 in human, mouse, rat, rabbit, hamster, canine, swine, and feline samples. MA1-06314 has sucessfully been used in Western blotting, flow cytometry, immunocytochemistry and immunohistochemistry applications. The MA1-06314 immunogens are cytokeratins from a human lung cancer cell line (MR21). Store at 4ºC or in small aliquots at -20ºC.

Cytokeratins are a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. They range in molecular weight between ~40 kDa and ~68 kDa and isoelectric pH between 4.9 - 7.8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 5/8
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone RCK102
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene KRT5
Gene Accession No. P05787, P11679, P13647, Q10758, Q6P6Q2, Q922U2
Gene Alias 3300001P10Rik; 58 kDa cytokeratin; AA960620; AL022697; AU019895; AW146334; Card2; CK5; CK-5; CK8; CK-8; CYK8; CYKER; cytokeratin 5; cytokeratin 8; cytokeratin 8 (370 AA); cytokeratin endo A; cytokeratin type II, component III; cytokeratin-5; Cytokeratin-8; cytokeratin-A; DDD; DDD1; EBS2; EndoA; epidermolysis bullosa simplex 2 Dowling-Meara/Kobner/Weber-Cockayne types; K2C8; K5; K8; KERA; keratin 5; keratin 5 (epidermolysis bullosa simplex Dowling-Meara/Kobner/Weber-Cockayne types); keratin 5 (epidermolysis bullosa simplex, Dowling-Meara/Kobner/Weber-Cockayne types); keratin 5, type II; keratin 8; keratin 8, type II; keratin complex 2, basic, gene 5; keratin complex 2, basic, gene 8; keratin, type II cytoskeletal 5; keratin, type II cytoskeletal 60 kDa, component III; Keratin, type II cytoskeletal 8; keratin, type II cytoskeletal 8-like protein; keratin-5; Keratin-8; KO; Krt-2.8; Krt2-5; Krt2-8; krt5; KRT5A; Krt8; Sak57; Tfip8; tuftelin interacting protein 8; type II cytokeratin A (no.8); type-II keratin Kb5; Type-II keratin Kb8
Gene Symbols KRT5, KRT8
Host Species Mouse
Immunogen Cytokeratins from human lung cancer cell line MR21.
Purification Method Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100152077, 100348891, 100352658, 100511564, 100687760, 100759488, 101081312, 101100735, 110308, 16691, 25626, 369017, 3852, 3856, 486513
Target Species Human, Mouse, Rat, Canine, Rabbit, Hamster, Pig, Feline
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.