Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 5/8 Mouse anti-Human, Clone: C50, Novus Biologicals™

Mouse Monoclonal Antibody
$219.50 - $487.50
Specifications
Antigen | Cytokeratin 5/8 |
---|---|
Clone | C50 |
Dilution | Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) |
Classification | Monoclonal |
Description
Cytokeratin 5/8 Monoclonal antibody specifically detects Cytokeratin 5/8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen)Specifications
Cytokeratin 5/8 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen | |
Monoclonal | |
Purified | |
RUO | |
Human | |
cell proliferation-inducing protein 46, CK18, CK5, CYK18, DDD, K18, K1CO, K5, Keratin, Keratin 18, Keratin 5, KRT18, krt5, KRT5A, KRTB, type I cytoskeletal 18, type II cytoskeletal 5 | |
Hybridoma produced by the fusion of splenocytes from mice immunized with cytoskeleton preparation of HeLa cells and mouse myeloma cells. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
C50 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) | |
Unconjugated | |
Mouse | |
Cell Biology, Cellular Markers | |
PBS | |
3852 | |
IgG1 | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title