Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 5 Antibody (104), Alexa Fluor™ 488

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289217AF488
Description
Cytokeratin 5 Monoclonal specifically detects Cytokeratin 5 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Cytokeratin 5 | |
Monoclonal | |
Alexa Fluor 488 | |
58 kDa cytokeratin, CK5, CK-5, cytokeratin-5, DDD, EBS2, epidermolysis bullosa simplex 2 Dowling-Meara/Kobner/Weber-Cockayne types, K5, keratin 5, keratin 5 (epidermolysis bullosa simplex, Dowling-Meara/Kobner/Weber-Cockaynetypes), keratin, type II cytoskeletal 5, Keratin-5, KRT5A, Type-II keratin Kb5 | |
This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the center region of the human Cytokeratin 5. | |
0.1 mL | |
Cell Biology, Cellular Markers | |
3852 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
104 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction