Learn More
Cytokeratin 6 Monoclonal Antibody (LHK6), Abbomax
Mouse Monoclonal Antibody
Supplier: Abbomax Inc 605612

Description
Positive control: Esophgus Cellular location: Cytoplasm.
Cytokeratin 6 Monoclonal antibody specifically detects Cytokeratin 6 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
Cytokeratin 6 | |
Monoclonal | |
0.5 mg/mL | |
PBS with proprietary stabilizer and 0.01% sodium azide | |
P02538, P04259, P48668, P50446, Q9Z331 | |
Mouse | |
Protein A | |
RUO | |
16687, 16688, 286887, 3853, 3854 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
LHK6 | |
Unconjugated | |
KRT6B | |
2310016L08Rik; 60-kDa keratin; BB144589; CK6A; CK-6A; CK6B; CK-6B; CK6C; CK-6C; CK6D; CK-6D; CK-6E; cytokeratin 6A; cytokeratin 6B; cytokeratin 6C; cytokeratin 6D; cytokeratin-6A; Cytokeratin-6B; cytokeratin-6C; Cytokeratin-6D; Cytokeratin-6E; Hom s 5; K6A; K6a keratin; K6B; K6C; K6D; K6E; kamp-keratin derived antimicrobial peptide; KDAMP; Ker2; keratin 6A; keratin 6A, , type II; keratin 6A, type II; keratin 6B; keratin 6B, type II; keratin 6C; keratin 6C, type II; keratin 6E; keratin complex 2, basic, gene 6a; keratin complex 2, basic, gene 6b; keratin complex 2, basic, gene 6c; keratin complex 2, gene 6a; keratin K6; keratin K6h; keratin, epidermal type II, K6A; keratin, epidermal, type II, K6B; keratin, type II cytoskeletal 6A; Keratin, type II cytoskeletal 6B; keratin, type II cytoskeletal 6C; keratin-6A; Keratin-6-alpha; Keratin-6B; keratin-6-beta; keratin-6C; keratin-like 1 (a type II keratin sequence); Krt2-6; Krt2-6a; Krt2-6b; Krt2-6c; Krt6; Krt6a; Krt6b; KRT6C; KRT6D; KRT6E; KRTL1; LOC683313; mK6[a]; mK6[b]; MK6a; mK6-alpha; mK6-beta; PC2; PC3; PC4; PPKNEFD; similar to keratin complex 2, basic, gene 6a; type-II keratin Kb10; Type-II keratin Kb12; type-II keratin Kb6 | |
A synthetic peptide of human Cytokeratin 6 | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.