Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytokeratin 6C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA580764
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA580764 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA580764 Supplier Invitrogen™ Supplier No. PA580764
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody has specificity for Human KRT6C.

KRT6C (Keratin 6C) is a Protein Coding gene. Diseases associated with KRT6C include Palmoplantar Keratoderma, Nonepidermolytic, Focal Or Diffuse and Focal Palmoplantar Keratoderma. Among its related pathways are Cytoskeletal Signaling and Cytoskeleton remodeling Neurofilaments. Gene Ontology (GO) annotations related to this gene include structural molecule activity. An important paralog of this gene is KRT6B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 6C
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene KRT6C
Gene Accession No. P48668
Gene Alias Cytokeratin 6C; KRT6C
Gene Symbols KRT6C
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the center region of the Human KRT6C.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286887
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.