Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 6a Antibody (KRT6/3997R), Alexa Fluor™ 532, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP314107AF532
This item is not returnable.
View return policy
Description
Cytokeratin 6a Monoclonal antibody specifically detects Cytokeratin 6a in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| Cytokeratin 6a | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| KRT6/3997R | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| Allergen Hom s 5, CK6A, CK-6A, CK6C, CK6D, CK-6D, cytokeratin 6A, cytokeratin 6C, cytokeratin 6D, Cytokeratin-6A, Cytokeratin-6D, K6A, K6C, K6D, keratin 6A, keratin 6C, keratin 6D, keratin, epidermal type II, K6A, keratin, type II cytoskeletal 6A, Keratin-6A, KRT6C, KRT6D, Type-II keratin Kb6 | |
| Recombinant fragment of human Cytokeratin 6a (exact sequence is proprietary) (Uniprot: P02538) | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, Stem Cell Markers | |
| 3853 | |
| Store at 4°C in the dark. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction