Learn More
Cytokeratin 6a Antibody (KRT6/3997R), Alexa Fluor™ 700, R&D Systems™
Shop All Novus Biologicals Products

Description
Specifications
Specifications
| Antigen | Cytokeratin 6a |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | KRT6/3997R |
| Conjugate | Alexa Fluor 700 |
| Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Allergen Hom s 5, CK6A, CK-6A, CK6C, CK6D, CK-6D, cytokeratin 6A, cytokeratin 6C, cytokeratin 6D, Cytokeratin-6A, Cytokeratin-6D, K6A, K6C, K6D, keratin 6A, keratin 6C, keratin 6D, keratin, epidermal type II, K6A, keratin, type II cytoskeletal 6A, Keratin-6A, KRT6C, KRT6D, Type-II keratin Kb6 |
| Host Species | Rabbit |
| Immunogen | Recombinant fragment of human Cytokeratin 6a (exact sequence is proprietary) (Uniprot: P02538) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.