Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 6a Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$231.00 - $262.00
Specifications
Antigen | Cytokeratin 6a |
---|---|
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Cytokeratin 6a Polyclonal antibody specifically detects Cytokeratin 6a in Human samples. It is validated for Western BlotSpecifications
Cytokeratin 6a | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cell Biology, Cellular Markers, Stem Cell Markers | |
PBS (pH 7.0) | |
3853 | |
IgG | |
Antigen and protein A Affinity-purified |
Western Blot 1:500-1:2000 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
Allergen Hom s 5, CK6A, CK-6A, CK6C, CK6D, CK-6D, cytokeratin 6A, cytokeratin 6C, cytokeratin 6D, Cytokeratin-6A, Cytokeratin-6D, K6A, K6C, K6D, keratin 6A, keratin 6C, keratin 6D, keratin, epidermal type II, K6A, keratin, type II cytoskeletal 6A, Keratin-6A, KRT6C, KRT6D, Type-II keratin Kb6 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Cytokeratin 6a. | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title