Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 6a, Mouse anti-Human, Clone: KRT6A/2368, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00 - $524.00
Specifications
Antigen | Cytokeratin 6a |
---|---|
Clone | KRT6A/2368 |
Concentration | 0.2 mg/mL |
Dilution | Flow Cytometry 1-2 ug/million cells, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array |
Description
Cytokeratin 6a Monoclonal specifically detects Cytokeratin 6a in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
Cytokeratin 6a | |
0.2 mg/mL | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
Unconjugated | |
Mouse | |
Cell Biology, Cellular Markers, Stem Cell Markers | |
10 mM PBS with 0.05% BSA | |
Allergen Hom s 5, CK6A, CK-6A, CK6C, CK6D, CK-6D, cytokeratin 6A, cytokeratin 6C, cytokeratin 6D, Cytokeratin-6A, Cytokeratin-6D, K6A, K6C, K6D, keratin 6A, keratin 6C, keratin 6D, keratin, epidermal type II, K6A, keratin, type II cytoskeletal 6A, Keratin-6A, KRT6C, KRT6D, Type-II keratin Kb6 | |
KRT6A | |
IgG2a κ | |
Protein A or G purified | |
This monoclonal antibody recognizes a protein of 56kDa, identified as Cytokeratin 6a (KRT6A). |
KRT6A/2368 | |
Flow Cytometry 1-2 ug/million cells, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
Human | |
P02538 | |
3853 | |
Recombinant full-length human Cytokeratin 6a protein | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title