Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 6a Mouse anti-Human, Clone: rKRT6A/2100, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00
Specifications
Antigen | Cytokeratin 6a |
---|---|
Clone | rKRT6A/2100 |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
Applications | Immunohistochemistry (Paraffin) |
Description
Cytokeratin 6a Monoclonal specifically detects Cytokeratin 6a in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
Cytokeratin 6a | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cell Biology, Cellular Markers, Stem Cell Markers | |
10 mM PBS with 0.05% BSA | |
3853 | |
IgG1 κ | |
Protein A or G purified |
rKRT6A/2100 | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Allergen Hom s 5, CK6A, CK-6A, CK6C, CK6D, CK-6D, cytokeratin 6A, cytokeratin 6C, cytokeratin 6D, Cytokeratin-6A, Cytokeratin-6D, K6A, K6C, K6D, keratin 6A, keratin 6C, keratin 6D, keratin, epidermal type II, K6A, keratin, type II cytoskeletal 6A, Keratin-6A, KRT6C, KRT6D, Type-II keratin Kb6 | |
Recombinant full-length human Cytokeratin 6a (KRT6A) protein | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title