Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytokeratin 6A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597910
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597910 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597910 Supplier Invitrogen™ Supplier No. PA597910
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cytokeratin 6A Polyclonal Antibody for Western Blot, IHC (P), ELISA

Keratins 6 and 16 are expressed in keratinocytes, which are undergoing rapid turnover in the suprabasal region (also known as hyperproliferation-related keratins). Keratin 6 is found in hair follicles, suprabasal cells of a variety of internal stratified epithelia, in epidermis, in both normal and hyperproliferative situations. Epidermal injury results in activation of keratinocytes which express CK6 and CK16. CK6 is strongly expressed in about 75% of head and neck squamous cell carcinomas. Expression of CK6 is particularly associated with differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 6A
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 4.69 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene KRT6A
Gene Accession No. P02538
Gene Alias 2310016L08Rik; 60-kDa keratin; CK6A; CK-6A; CK6C; CK-6C; CK6D; CK-6D; CK-6E; cytokeratin 6A; cytokeratin 6C; cytokeratin 6D; cytokeratin-6A; cytokeratin-6C; Cytokeratin-6D; cytokeratin-6E; Hom s 5; K6A; K6a keratin; K6C; K6D; kamp-keratin derived antimicrobial peptide; KDAMP; Ker2; keratin 6A; keratin 6A, , type II; keratin 6A, type II; keratin complex 2, basic, gene 6a; keratin complex 2, basic, gene 6c; keratin complex 2, gene 6a; keratin K6; keratin K6h; keratin, epidermal type II, K6A; keratin, type II cytoskeletal 6A; keratin, type II cytoskeletal 6C; Keratin-6A; keratin-6-alpha; keratin-6C; Krt2-6; Krt2-6a; Krt2-6c; Krt6; KRT6A; KRT6C; KRT6D; LOC683313; mK6[a]; MK6a; mK6-alpha; PC3; similar to keratin complex 2, basic, gene 6a; type-II keratin Kb12; type-II keratin Kb6
Gene Symbols KRT6A
Host Species Rabbit
Immunogen Recombinant Human Keratin, type II cytoskeletal 6A protein (2-564AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3853
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.