Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytokeratin 6B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112594
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112594 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112594 Supplier Invitrogen™ Supplier No. PA5112594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cytokeratin 6B Polyclonal Antibody for Western Blot, ELISA

The protein encoded by this gene is a member of the keratin gene family. The type II cytokeratins consist of basic or neutral proteins which are arranged in pairs of heterotypic keratin chains coexpressed during differentiation of simple and stratified epithelial tissues. As many as six of this type II cytokeratin (KRT6) have been identified; the multiplicity of the genes is attributed to successive gene duplication events. The genes are expressed with family members KRT16 and/or KRT17 in the filiform papillae of the tongue, the stratified epithelial lining of oral mucosa and esophagus, the outer root sheath of hair follicles, and the glandular epithelia. Mutations in these genes have been associated with pachyonychia congenita. The type II cytokeratins are clustered in a region of chromosome 12q12-q13.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 6B
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.01 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene KRT6B
Gene Accession No. P04259
Gene Alias BB144589; CK6B; CK-6B; cytokeratin 6B; cytokeratin-6B; K6B; keratin 6B; keratin 6B, type II; keratin complex 2, basic, gene 6b; keratin, epidermal, type II, K6B; keratin, type II cytoskeletal 6B; keratin-6B; keratin-6-beta; keratin-like 1 (a type II keratin sequence); Krt2-6b; KRT6B; KRTL1; mK6[b]; mK6-beta; PC2; PC4; type-II keratin Kb10
Gene Symbols KRT6B
Host Species Rabbit
Immunogen Recombinant Human Keratin, type II cytoskeletal 6B protein (1-280AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3854
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.