Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 7/17 Antibody (C-46), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308343MFV450
Description
Cytokeratin 7/17 Monoclonal antibody specifically detects Cytokeratin 7/17 in Human, Bovine, Ferret (Negative), Mouse (Negative), Rabbit (Negative), Rat (Negative) samples. It is validated for Flow CytometrySpecifications
Cytokeratin 7/17 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Bovine, Ferret (Negative), Mouse (Negative), Rabbit (Negative), Rat (Negative) | |
Purified |
Flow Cytometry | |
C-46 | |
Flow Cytometry | |
CK7, CK-7, cytokeratin 7, Cytokeratin-7, K2C7, K7keratin, 55K type II cytoskeletal, keratin 7, keratin, type II cytoskeletal 7, keratin-7, MGC129731, MGC3625, Sarcolectin, SCLkeratin, simple epithelial type I, K7, type II mesothelial keratin K7, Type-II keratin Kb7 | |
Cytoskeletal preparation from HeLa cells (human cervical carcinoma cells) | |
0.1 mL | |
Cell Biology, Cellular Markers | |
3855 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction