Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 7/17 Mouse anti-Human, Clone: C46, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311896100UG
Description
Cytokeratin 7/17 Monoclonal antibody specifically detects Cytokeratin 7/17 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen)Specifications
Cytokeratin 7/17 | |
Monoclonal | |
Unconjugated | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) | |
C46 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen | |
CK7, CK-7, cytokeratin 7, Cytokeratin-7, K2C7, K7keratin, 55K type II cytoskeletal, keratin 7, keratin, type II cytoskeletal 7, keratin-7, MGC129731, MGC3625, Sarcolectin, SCLkeratin, simple epithelial type I, K7, type II mesothelial keratin K7, Type-II keratin Kb7 | |
Hybridoma produced by the fusion of splenocytes from mice immunized with cytoskeleton preparation from HeLa cells and mouse myeloma cells. | |
100 μg | |
Cell Biology, Cellular Markers | |
3855 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction