Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 7, 8 Antibody (LDS68) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10
Specifications
Antigen | Cytokeratin 7, 8 |
---|---|
Clone | LDS68 |
Concentration | 1 mg/mL |
Dilution | Immunohistochemistry 1:10 - 1:500 |
Applications | Immunohistochemistry |
Description
Cytokeratin 7, 8 Monoclonal antibody specifically detects Cytokeratin 7, 8 in Human samples. It is validated for ImmunohistochemistrySpecifications
Cytokeratin 7, 8 | |
1 mg/mL | |
Immunohistochemistry | |
Unconjugated | |
Mouse | |
Cellular Signaling | |
PBS | |
3855 | |
IgG1 | |
Protein A purified |
LDS68 | |
Immunohistochemistry 1:10 - 1:500 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CK7, CK-7, CK8, CK-8, Cytokeratin-7, Cytokeratin-8, keratin 8, keratin, type II cytoskeletal 7, keratin-7 | |
Cytoskeletons of the human urothelial RT4 cell line. | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title