Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 7, 8 Antibody (LDS68), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250460G
Description
Cytokeratin 7, 8 Monoclonal specifically detects Cytokeratin 7, 8 in Human samples. It is validated for Immunohistochemistry.Specifications
Cytokeratin 7, 8 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Cytoskeletons of the human urothelial RT4 cell line. | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. |
Immunohistochemistry | |
LDS68 | |
Immunohistochemistry | |
Mouse | |
Protein A purified | |
Cellular Signaling | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction