Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 7 Antibody (KRT7/1198) - IHC-Prediluted, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP248145
Description
Cytokeratin 7 Monoclonal specifically detects Cytokeratin 7 in Human, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Cytokeratin 7 | |
Monoclonal | |
Unconjugated | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
KRT7 | |
Recombinant full-length human Cytokeratin 7 protein (Uniprot: P08729) | |
Protein A or G purified | |
RUO | |
Primary | |
It recognizes an intermediate filament protein (IFP) of 55kDa, which is identified as cytokeratin 7. This monoclonal antibody is highly specific to cytokeratin 7 and shows no cross-reaction with other IFPs. Cytokeratin 7 is a basic cytokeratin, which is found in most glandular and transitional epithelia but not in the stratified squamous epithelia. Keratin 7 is expressed in the epithelial cells of ovary, lung, and breast but not of colon, prostate, or gastrointestinal tract. This monoclonal antibody is highly useful in distinguishing ovarian carcinomas (keratin 7+) from colon carcinomas (keratin 7-). | |
Store at 4C. | |
IgG1 κ |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
KRT7/1198 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
CK7, CK-7, cytokeratin 7, Cytokeratin-7, K2C7, K7keratin, 55K type II cytoskeletal, keratin 7, keratin, type II cytoskeletal 7, keratin-7, MGC129731, MGC3625, Sarcolectin, SCLkeratin, simple epithelial type I, K7, type II mesothelial keratin K7, Type-II keratin Kb7 | |
Mouse | |
55 kDa | |
7 mL | |
Cytoskeleton Markers | |
3855 | |
Human, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction