Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Cytokeratin 7 Antibody (KRT7/760 + OV-TL12/30), Allophycocyanin, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP247943APC
Description
Cytokeratin 7 Monoclonal specifically detects Cytokeratin 7 in Human, Rat samples. It is validated for Flow Cytometry.Specifications
| Cytokeratin 7 | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| KRT7 | |
| Recombinant full-length human KRT7 protein (KRT7/760); OTN 11, ovarian carcinoma cell line (OV-TL12/30) (Uniprot: P08729) | |
| 0.1 mL | |
| Primary | |
| It recognizes an intermediate filament protein (IFP) of 55kDa, which is identified as cytokeratin 7. This monoclonal antibody is highly specific to cytokeratin 7 and shows no cross-reaction with other IFPs. Cytokeratin 7 is a basic cytokeratin, which is found in most glandular and transitional epithelia but not in the stratified squamous epithelia. Keratin 7 is expressed in the epithelial cells of ovary, lung, and breast but not of colon, prostate, or gastrointestinal tract. This monoclonal antibody is highly useful in distinguishing ovarian carcinomas (keratin 7+) from colon carcinomas (keratin 7-). | |
| Store at 4C in the dark. | |
| IgG1 κ | 
| Flow Cytometry | |
| KRT7/760 + OV-TL12/30 | |
| Flow Cytometry | |
| CK7, CK-7, cytokeratin 7, Cytokeratin-7, K2C7, K7keratin, 55K type II cytoskeletal, keratin 7, keratin, type II cytoskeletal 7, keratin-7, MGC129731, MGC3625, Sarcolectin, SCLkeratin, simple epithelial type I, K7, type II mesothelial keratin K7, Type-II keratin Kb7 | |
| Mouse | |
| Protein A or G purified | |
| Cytoskeleton Markers | |
| 3855 | |
| Human, Rat | |
| Purified | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            