Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 7 Antibody (KRT7/903), Novus Biologicals™

Mouse Monoclonal Antibody
$268.00 - $524.00
Specifications
| Antigen | Cytokeratin 7 |
|---|---|
| Clone | KRT7/903 |
| Concentration | 0.2mg/mL |
| Dilution | Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml, SDS-Page, Immunofluorescence 0.5 - 1.0 ug/ml |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), SDS-Page, Immunofluorescence |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP24481700
![]() |
Novus Biologicals
NBP2448170.02MG |
0.02 mg |
Each for $268.00
|
|
|||||
NBP24481701
![]() |
Novus Biologicals
NBP2448170.1MG |
0.1 mg |
Each for $524.00
|
|
|||||
NBP24481702
![]() |
Novus Biologicals
NBP2448170.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
Cytokeratin 7 Monoclonal specifically detects Cytokeratin 7 in Human, Ferret (Negative), Mouse (Negative), Rabbit (Negative), Rat (Negative) samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Cytokeratin 7 | |
| 0.2mg/mL | |
| Flow Cytometry, Immunohistochemistry (Paraffin), SDS-Page, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cytoskeleton Markers | |
| 10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| 3855 | |
| Recombinant full-length human KRT7 protein | |
| Primary | |
| Store at 4C. | |
| 54 kDa |
| KRT7/903 | |
| Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml, SDS-Page, Immunofluorescence 0.5 - 1.0 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Ferret (Negative), Mouse (Negative), Rabbit (Negative), Rat (Negative) | |
| CK7, CK-7, cytokeratin 7, Cytokeratin-7, K2C7, K7keratin, 55K type II cytoskeletal, keratin 7, keratin, type II cytoskeletal 7, keratin-7, MGC129731, MGC3625, Sarcolectin, SCLkeratin, simple epithelial type I, K7, type II mesothelial keratin K7, Type-II keratin Kb7 | |
| KRT7 | |
| IgG1 | |
| Protein A or G purified | |
| Cytokeratin 7 is a basic cytokeratin, which is found in most glandular and transitional epithelia but not in the stratified squamous epithelia. Keratin 7 is expressed in the epithelial cells of ovary, lung, and breast but not of colon, prostate, or gastrointestinal tract. Antibody to cytokeratin is useful in distinguishing ovarian carcinomas (keratin 7+) from colon carcinomas (keratin 7-). |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title