Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 7 Antibody (rKRT7/8763), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP320235100UG
Description
Cytokeratin 7 Monoclonal antibody specifically detects Cytokeratin 7 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Cytokeratin 7 | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 μg/mL | |
CK7, CK-7, cytokeratin 7, Cytokeratin-7, K2C7, K7keratin, 55K type II cytoskeletal, keratin 7, keratin, type II cytoskeletal 7, keratin-7, MGC129731, MGC3625, Sarcolectin, SCLkeratin, simple epithelial type I, K7, type II mesothelial keratin K7, Type-II keratin Kb7 | |
Recombinant full-length human Cytokeratin 7 protein | |
100 μg | |
Cell Biology, Cellular Markers | |
3855 | |
Store at 4&drg;C. Do not freeze. | |
IgG2b κ |
Immunohistochemistry (Paraffin) | |
rKRT7/8763 | |
Unconjugated | |
10mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction