Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 7 Antibody (SPM270), DyLight 488 , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311594G
Description
Cytokeratin 7 Monoclonal antibody specifically detects Cytokeratin 7 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Cytokeratin 7 | |
Monoclonal | |
DyLight 488 | |
CK7, CK-7, cytokeratin 7, Cytokeratin-7, K2C7, K7keratin, 55K type II cytoskeletal, keratin 7, keratin, type II cytoskeletal 7, keratin-7, MGC129731, MGC3625, Sarcolectin, SCLkeratin, simple epithelial type I, K7, type II mesothelial keratin K7, Type-II keratin Kb7 | |
OTN 11, ovarian carcinoma cell line | |
0.1 mL | |
Cell Biology, Cellular Markers | |
3855 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
SPM270 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction