Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 7/Keratin K7 Mouse anti-Human, Clone: OVTL12/30, MUbio
SDP

Catalog No. 502101034
Encompass
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-210-1034 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-210-1034 Supplier Vector Laboratories Supplier No. MUB0316P
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MKs20-8 may be used as a marker for certain types of carcinomas such as adenocarcinomas of the colon, transitional cell carcinomas of the bladder and Merkel cell tumors of the skin; it may also be used as a sensitive detection of intestinal and gastric foveolar epithelium, urothelial umbrella cells, Merkel cells of epidermis as well as tumors originating therefrom (e.g. primary and metastatic colorectal carcinoma). The antibody may be stored at 4 C. For prolonged storage prepare appropriate aliquots and store at or below -20 C. Prior to use, an aliquot is thawed slowly in the dark at ambient temperature, spun down again and used to prepare working dilutions by adding sterile phosphate buffered saline (PBS, pH 7.2). Repeated thawing and freezing should be avoided. Working dilutions should be stored at 4 C, not refrozen, and preferably used the same day. If a slight precipitation occurs upon storage, this should be removed by centrifugation. It will not affect the performance or the concentration of the product.

Cytokeratin 7/Keratin K7 Monoclonal antibody specifically detects Cytokeratin 7/Keratin K7 in Human samples. It is validated for Flow Cytometry, Immunoblot, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen Cytokeratin 7/Keratin K7
Applications Flow Cytometry, Immunoblot, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OVTL12/30
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH
Gene KRT7
Gene Accession No. P08729
Gene Alias C SCL; CK7; CK-7; Cytokeratin 7; cytokeratin-7; D15Wsu77e; K2C7; K7; Kb7; keratin 7; keratin 7, type II; keratin associated protein 7.1; keratin complex 2, basic, gene 7; keratin, 55K type II cytoskeletal; keratin, simple epithelial type I, K7; keratin, type II cytoskeletal 7; Keratin-7; keratin-associated protein 7-1; keratin-associated protein 7-1-like protein; Krt2-7; KRT7; krt7 {ECO:0000250; LOC100755511; LOC100861181; mgC129731; mgC3625; sarcolectin; SCL; type II keratin Kb7; type II mesothelial keratin K7; type-II keratin Kb7; UniProtKB:P08729}
Host Species Mouse
Immunogen Cytokeratin 7 / Keratin K7
Purification Method Protein A/G
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3855
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.