Learn More
Invitrogen™ Cytokeratin 7 Monoclonal Antibody (OVTL12/30)

Description
MA1-06316 detects Cytokeratin 7 in human samples. MA1-06316 has sucessfully been used in Western blotting, immunocytochemistry, flow cytometry, and immunohistochemistry. Pretreatment is required for paraffin sections; 0.1% pronase/0.1% trypsine in PBS or 0.5% pepsine in HCL or microwave pretreatment suggested. The MA1-06316 immunogen is a cytoskeletal preparation of OTN-11 ovarian carcinoma cell line. Store at 4°C, or in small aliquots at -20°C.
Specifications
Specifications
| Antigen | Cytokeratin 7 |
| Applications | Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OVTL12/30 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide |
| Gene | KRT7 |
| Gene Accession No. | P08729 |
| Gene Alias | CK7; CK-7; Cytokeratin 7; cytokeratin-7; D15Wsu77e; K2C7; K7; Kb7; keratin 7; keratin 7, type II; keratin associated protein 7.1; keratin complex 2, basic, gene 7; keratin, 55K type II cytoskeletal; keratin, simple epithelial type I, K7; keratin, type II cytoskeletal 7; Keratin-7; keratin-associated protein 7-1; keratin-associated protein 7-1-like protein; Krt2-7; KRT7; krt7 {ECO:0000250; LOC100755511; LOC100861181; NACHT, LRR and PYD domains-containing protein 3; sarcolectin; SCL; si:ch73-233m11.1; si:ch73-233m11.2; similar to keratin 7; type II keratin Kb7; type II mesothelial keratin K7; type-II keratin Kb7; UniProtKB:P08729} |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.