Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 7 Rabbit anti-Human, Clone: 6E6, Bioss
SDP

Catalog No. 501989561
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9561 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9561 Supplier Bioss Supplier No. BSM52065R
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Cytokeratin 7 blocks interferon-dependent interphase and stimulates DNA synthesis in cells. Involved in the translational regulation of the human papillomavirus type 16 E7 mRNA (HPV16 E7).

Specifications

Antigen Cytokeratin 7
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone 6E6
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 40% glycerol and 0.05% sodium azide; pH
Gene KRT7
Gene Accession No. P08729
Gene Alias C SCL; CK7; CK-7; Cytokeratin 7; Cytokeratin-7; D15Wsu77e; K2C7; K7; Kb7; keratin 7; keratin 7, type II; keratin associated protein 7.1; keratin complex 2, basic, gene 7; keratin, 55K type II cytoskeletal; keratin, simple epithelial type I, K7; Keratin, type II cytoskeletal 7; Keratin-7; keratin-associated protein 7-1; keratin-associated protein 7-1-like protein; Krt2-7; KRT7; krt7 {ECO:0000250; LOC100755511; LOC100861181; MGC129731; MGC3625; Sarcolectin; SCL; type II keratin Kb7; type II mesothelial keratin K7; Type-II keratin Kb7; UniProtKB:P08729}
Gene Symbols KRT7
Host Species Rabbit
Immunogen Recombinant human Cytokeratin 7 protein, around C-terminal 150aa.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3855
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.