Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 7 Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502206540
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-6540 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-6540 Supplier Fabgennix Inc Supplier No. CK7701AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cytokeratin 7 Polyclonal antibody specifically detects Cytokeratin 7 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen Cytokeratin 7
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene KRT7
Gene Accession No. P08729, Q6IG12, Q9DCV7
Gene Alias C SCL; CK7; CK-7; Cytokeratin 7; cytokeratin-7; D15Wsu77e; K2C7; K7; Kb7; keratin 7; keratin 7, type II; keratin associated protein 7.1; keratin complex 2, basic, gene 7; keratin, 55K type II cytoskeletal; keratin, simple epithelial type I, K7; keratin, type II cytoskeletal 7; Keratin-7; keratin-associated protein 7-1; keratin-associated protein 7-1-like protein; Krt2-7; KRT7; krt7 {ECO:0000250; LOC100755511; LOC100861181; MGC129731; MGC3625; NACHT, LRR and PYD domains-containing protein 3; sarcolectin; SCL; si:ch73-233m11.1; si:ch73-233m11.2; similar to keratin 7; type II keratin Kb7; type II mesothelial keratin K7; type-II keratin Kb7; UniProtKB:P08729}
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 10-60 on human keratin, type II cytoskeletal 7 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110310, 300242, 3855
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.